Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silver lactate, powder

Supplier: Electron Microscopy Sciences
Warning
Synonyms: Lactic acid silver salt

21000
100504-262EA 60.64 USD InStock
100504-262
Silver lactate, powder
Silver lactate
Used to localize intracellular anions (Cl⁻).

Rference: Komnick, (1962), Elektronemikrokopische Lokalisation von Na+ und Cl in Zellan und Geweban. Protoplasma, 55,414.


Formula: C₃H₅AgO₃
MW: 196.938 g/mol
Melting Pt: 122 °C (252 °F)
MDL Number: MFCD00043279
CAS Number: 15768-18-0
https://us-legacy.vwr.com/stibo/web/std.lang.all/48/93/28984893.jpg
Order Now
Specification Test Results
Silver content Approx. 90% min

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty