Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium carbonate, anhydrous ACS

Supplier: Electron Microscopy Sciences
Warning
Synonyms: Na-XCarbonic acid sodium saltDisodium carbonateSodaBisodium carbonateCarbonic acid, disodium saltSoda ash

21135
100504-276EA 86.62 USD InStock
100504-276
Sodium carbonate, anhydrous ACS
Sodium carbonate


Formula: Na₂CO₃
MW: 105.99 g/mol
Boiling Pt: 1600 °C (2912 °F)
Melting Pt: 851 °C (1564 °F)
Density: 2.532 g/cm³ (21.13 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/48/98/28984898.jpg
Order Now
Specification Test Results
Ammonium hydroxide ppt 0.01%
Arsenic 1 ppm
Ca and Mg 0.01%
Chloride 0.001%
Heavy metals 5 ppm
Insoluble matters 0.01%
Iron 5 ppm
Loss on heating (280 °C) 1.0%
Nitrogen compounds 0.001%
Phosphate 0.001%
Potassium 0.005%
Silica (SiO₂) 0.005%
Sulfur compounds (as S) 0.003%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty