Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Toluene ≥99.9% ACS

Supplier: Electron Microscopy Sciences
Danger
Synonyms: MethacideBenzene, methylMethylbenzolToluolPhenylmethaneMethylbenzeneMethane, phenyl-

22030 22032
100504-320EA 20.37 USD InStock
100504-320 100504-322
Toluene ≥99.9% ACS
Toluene


Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 110…111 °C (230…232 °F)
Melting Pt: –95 °C (–139 °F)
Density: 0.87 g/cm³ (7.26 lbs/gal.)
Flash Pt: 4 °C (39 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
UN: 1294
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/49/21/28984921.jpg
Order Now
Specification Test Results
Assay 99.9%
Residue after evaporation 0.001%
Specific gravity (H₂O=1) 0.866
Substance darkened by H₂SO₄ To pass test
Sulfur compounds (as S) 0.003%
Water 0.03%
Color (APHA) 10

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty