Would you like to log in again?
Mayer Mucicarmine Method, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
26320-04 26320-01 26320-03 26320-02
100504-588EA 123.5 USD InStock
100504-588 100504-886 100504-590 100504-592
Mayer Mucicarmine Method, Electron Microscopy Sciences
Stains Microbiology Stains
Mayer Mucicarmine Method for mucosubstances stains
Note: rose color due to carmin staining will be obscured if sections are overstained with Weigert’s Hematoxylin and/or Metanil Yellow solution.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty