Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminium oxide basic 90

Supplier: Electron Microscopy Sciences
Warning

10400
101410-620EA 137.72 USD InStock
101410-620
Aluminium oxide basic 90
Aluminium oxide basic
Aluminum oxide powder with 63 to 200 micron particles is ideal for use with the Technovit® 9100.


Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2980 ±60 °C (5396 ±140 °F)
Melting Pt: 2050 °C (3722 °F)
Density: 3,97 g/cm³ (33.13 lbs/gal.)
MDL Number: MFCD00003424
CAS Number: 1344-28-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty