Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydroquinone 1% in aqueous solution

Supplier: Electron Microscopy Sciences
Synonyms: 1,4-DihydroxybenzeneQuinol

16795-1
101410-696EA 98.26 USD InStock
101410-696
Hydroquinone 1% in aqueous solution
Hydroquinone
Hydroquinone Aqueous solution is available in concentrations of 1% aqueous, 2% aqueous, 4.0% aqueous.


Formula: C₆H₄(OH)₂
MW: 110.11 g/mol
MDL Number: MFCD00002339
CAS Number: 123-31-9
UN: 3077
ADR: 9,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/48/37/28984837.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty