Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phosphotungstic acid hydrate, crystals, high purity

Supplier: Electron Microscopy Sciences
Danger
Synonyms: Wolframophosphoric acid hydratePhosphotungstic acid hydrate12-Tungstophosphate hydrateDodecawolframophosphoric acid hydrateTungstophosphoric Acid Hydrate

19500
100504-194EA 125.77 USD InStock
100504-194
Phosphotungstic acid hydrate, crystals, high purity
Phosphotungstic acid hydrate
Phosphotungstic acid can be used as a fixative and widely used as a negative stain (Issidorides, M.R., and Kasorchis, T. (1981)).

Dispersed and compact chromatin demonstrated with a new EM method that is phosphotungstic acid-hematoxylin block-staining (Histochemistry 73,21).
As spray method, a negative staining-carbon film technique for studying viruses in the electron microscope (Horne, R.W., and Pasquali-Ronchette, I. (1974)).
Preparation procedures for examining icosahedral and filamentous viruses (J. Ultrastruct. Res. 47, 361. Locke, M., and Kirhman, N. (1971)).


Formula: H₃[P(W₃O₁₀)₄]·nH₂O
MW: 2898.07 g/mol
Melting Pt: 95 °C (203 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011342
CAS Number: 12501-23-4
UN: 3264
ADR: 8,II
Order Now
Specification Test Results
Ammonia (NH ) 0.004%
Chloride (Cl) 0.03%
Insoluble matter 0.02%
Nitrate (NO₃) 0.004%
Sulfate (SO₄) 0.005%
Heavy metals and iron No color formation on addition of hydrogen sulfide

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty