Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
NGAL (pig) ELISA kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
BPD-KIT-044
10049-718EA 1063.42 USD InStock
10049-718
NGAL (pig) ELISA kit, Enzo Life Sciences
Assays ELISAs
For the quantitative determination of NGAL in pig urine, plasma, serum, tissue extracts, or culture supernatants.
A colorimetric immunometric ELISA kit for the rapid measurement of NGAL in pig samples. Absorbance is read at 450nm.
Ordering information: Contains 1: Microwell plate (96 precoated wells); 2: 50ml 5x Sample Diluent Conc.; 3: 7 x 1ml  Pig NGAL Calibrator (1-7. 0, 10, 20, 40, 100, 200, 400 pg/mL); 4: 40ml 25x Wash Solution Conc.; 5: 12ml Biotinylated Pig-NGAL Antibody; 6: 12ml HRP-Streptavidin;  7:12ml TMB Substrate; and 8: 12ml Stop Solution.. Manufactured by BioPorto Diagnostics.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty