Would you like to log in again?
NGAL (rat) ELISA kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
BPD-KIT-046
10049-720EA 992.06 USD InStock
10049-720
NGAL (rat) ELISA kit, Enzo Life Sciences
Assays ELISAs
For the quantitative determination of NGAL in rat urine, plasma, serum, tissue extracts, or culture supernatants.
A colorimetric immunometric ELISA kit for the rapid measurement of NGAL in rat samples. Absorbance is read at 450nm.
Ordering information: Contains 1: Microwell plate coated with anti-NGAL (rat) antibody (96 wells) 2: 5X Sample Diluent 3: NGAL Rat Calibrators 1-8. 4: 25X Wash Solution Concentrate 5: Biotinylated Rat NGAL Antibody 6: HRP-Streptavidin 7: TMB Substrate 8: Stop Solution. Manufactured by BioPorto Diagnostics.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty