Would you like to log in again?
Bovine Desoxyribonuclease I (from Pancreas)
Supplier: EMD Millipore
Calbiochem®
260913-25MU 260913-10MU
80510-414EA 790.09 USD InStock
80510-414 80510-412
Bovine Desoxyribonuclease I (from Pancreas)
Enzymes
Native DNase I from bovine pancreas.
An endonuclease that preferentially cleaves phosphodiester linkages adjacent to pyrimidine nucleotides to yield 5′-phosphate terminated polynucleotides with a free hydroxyl group at the 3′ position. Acts on single and double stranded DNA. Note: 1 MU = 1,000,000 Dornase units.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty