Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Warning Voltage and Electrical Restricted Area Signs, National Marker
Supplier: National Marker
W411PB W410RB W410P ESW500AB WGA26AB WGA26RB W422RB ESW500AP WGA26PB W410PB W410R W411AB ESW500RB W422PB WGA22AP W422AB ESW500PB W411RB
94023-178EA 20.3 USD InStock
94023-178 94023-254 94013-706 94023-188 94023-176 94023-594 94023-186 94013-704 94023-252 94013-710 94023-180 94023-184 94023-250 94023-182 94013-708 94023-604 94023-608 94023-606
Warning Voltage and Electrical Restricted Area Signs, National Marker
Signs
Safety signs are the perfect way to alert your employees and guests when a hazard is present in your facility.
  • Ideal for indoor and outdoor labeling
  • Available in Alum, PS vinyl and Rigid plastic (thick polystyrene) materials
  • .040 alum is weatherproof, non-corrosive and has baked-on enamel on both sides
  • .004 PS vinyl material has pressure sensitive adhesive backing
  • .050 rigid plastic material has radius corners and pre-drilled corner mounting holes
Eliminate accidents and keep employees safe by ensuring all areas in the workplace are clearly marked for electrical hazards. These hazards can range from high voltage service (over 600 volts) to unexpected lines and cables, to access space for work or servicing, and locations to shut power off.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty