Would you like to log in again?
Hypersil GOLD™ aQ HPLC Cartridges and Columns, Thermo Scientific
Supplier: Thermo Scientific
25305-012101 25305-013001 25303-011001 25303-014001 25305-014001 25303-013001 25305-012106 25303-012101
10041-352EA 595.55 USD InStock
10041-352 10045-556 10041-354 10045-558 10045-560 10045-494 10045-562 10041-350
Hypersil GOLD™ aQ HPLC Cartridges and Columns, Thermo Scientific
Chromatography Columns
Achieve exceptional peak shape and resolution for your HPLC and LC/MS applications with Thermo Scientific Hypersil GOLD HPLC columns.
  • Ensures optimized peak shape, resolution, sensitivity and efficiency
  • Provides only controlled secondary interactions to ensure excellent peak shape for all analyte types
  • Polar endcapping of media also makes it usable in 100% aqueous mobile phases without the risk of loss of performance or poor stability
These endcapped, ultrapure, silica-based columns deliver significant reduction in peak tailing using generic gradients with C18 selectivity. With their excellent resolution, efficiency, and sensitivity, Hypersil GOLD columns give you confidence in the accuracy and quality of your analytical data. 1.9µm particle size columns can improve the speed and efficiency of your applications.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD Holders. Cartridges with a 1 mm Int.Ø require the 1 mm UNIGUARD Holder, 2.1/3 mm Int.Ø require the 2/3 mm UNIGUARD Holder and 4/4.6 mm Int.Ø require the 4/4.6 mm UNIGUARD Holder.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty