Would you like to log in again?
Eppendorf® epTIPS®, Biopur® Tips
Supplier: Eppendorf
epT.I.P.S.
022491113 022491083 022491148 0030075277 022491091 022491148 022491105 022491067 022491121 0030075269 0030075226 0030075285 022491164 0030075277 0030075242 0030075250 0030075234 022491130 022491156 022491156 022491130
47745-078CS 114.09 USD InStock
47745-078 CA47745-066 CA47745-078 47745-066 47745-068 CA47745-076 CA47745-068 CA47745-080 47745-070 47745-080 CA47745-070 47745-072 47745-082 47745-074 47747-990 89126-934 CA47745-074 CA89126-934 47745-076 CA47745-082 CA47745-072
Eppendorf® epTIPS®, Biopur® Tips
For highest purity demands in cell culture, nucleic acid analytics, or hygiene monitoring.
  • Continuous quality control of each batch by an independent laboratory
  • Optimal wetting properties
  • High transparency
  • Ergonomically optimized cone geometry
  • Commitment to both product and production quality
For epTIPS®, the original Eppendorf 'Totally Integrated Pipetting System', the tips are optimally coordinated to Eppendorf pipettors. This has resulted in minimal attachment and ejection forces with the highest level of tightness. epTIPS® can be used with pipettors from other manufacturers, as well.
Biopur® tips meet the highest demands of the clinical, pharmaceutical, cosmetic, food, and other industries. The purity is ensured by an elaborate, automated manufacturing process which safeguards the product against contamination with biological substances. Lot sepcific certificates provided by an external accredited laboratory confirm compliance with the following criteria: sterile, DNase-, RNase-, PCR inhibitor-, ATP-, Pyrogen-, human DNA- and bacterial DNA-free. These certificates are available upon request.
Certifications: Meet EN ISO 8655 requirements.
Packaging: Available as epTIPS® Singles (packaged individually) or as epTIPS® Racks (boxes containing racks of 96, 48, or 24 tips; depending on tip volume).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty