Would you like to log in again?
epTIPS® PCR Clean Tips, Eppendorf®
Supplier: Eppendorf
epT.I.P.S. epTIPS®
022491873 022491881 022491814 0030071662 022491865
47745-216CS 85 USD InStock
CA47745-202 CA47745-214 47745-216 CA47745-212 CA47745-216
epTIPS® PCR Clean Tips, Eppendorf®
Pipette Tips
The perfect choice for isolation and analytics (PCR/qPCR, microarrays) of RNA, DNA, and nucleic acids.
  • Free of DNase and RNase
  • PCR inhibitor free
  • Continuous quality control of each batch by an independent laboratory
For epTIPS® - the original Eppendorf 'Totally Integrated Pipetting System' - the tips are optimally coordinated to Eppendorf® pipettes. This results in minimal attachment and ejection forces with the highest level of tightness. epTIPS® pipette tips can be used with pipettes from other manufacturers. Optimal wetting properties, high transparency, and ergonomically optimized cone geometry are a testament to the commitment to product and production quality.
Eppendorf® PCR Clean consumables are certified free of DNases, RNases, human DNA, and PCR inhibitors. This purity grade is achieved by using pure raw materials, autmoated production under cleanroom conditions, quality and functional checks for all lots, and continuous quality assurance throughout the entire production process. With the strictest control criteria, internally and externally monitored, we guarentee consistent, high-quality pipette tips from lot to lot. Lot-specific certificates are available upon request and purity is certified by an independent, third party laboratory.
Certifications: Meet EN ISO 8655 requirements.
Packaging: Available as epTIPS® Racks (boxes containing racks of 96, 48, or 24 tips, depending on tip volume), or as epTIPS® Reloads (packaged as either dual-sided or stacked racks, depending on tip volume).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty