Would you like to log in again?
NPXO Series Powder Free Nitrile Exam Gloves, CT International
Supplier: CT International
NPXO103 NPXO101
33502-656CS 317.3 USD InStock
33502-656 33502-652
NPXO Series Powder Free Nitrile Exam Gloves, CT International
Gloves
NPXO Series powder free nitrile exam gloves are manufactured for outstanding strength, stretch and fit.
  • Ambidextrous, 10" length
  • Strong and durable
  • No latex proteins, tapered and beaded cuff
  • Textured finger tips and palm
They are 100% nitrile offer textured finger tips, have a super soft feel and come in orange color.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty