Would you like to log in again?
Lounsbury Placenta Curette, OR Grade, Sklar
Supplier: Sklar
90-5713
10194-220EA 236.66 USD InStock
10194-220
Lounsbury Placenta Curette, OR Grade, Sklar
Curettes
Lounsbury Placenta Curette is used to assist in removal of any remaining placenta.
The overall length of this instrument is 13". The handle is a rounded square with grooves for improved grip, along with a thumb/finger rest on two sides. The shaft is thin and round and leads to a large tear-drop shaped, fenestrated tip. The tip is approximately 1" long, 3/4" wide from side to side, and has a 1/4" thick wall. There is one sharp edge to assist in the removal procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty