Tips for all routine laboratory applications.
- Fine graduations make visual checks of pipetting volume even easier
- Elongated tips reach the base of narrow and high vessels without touching the top of the vessel edge
- Color-coded for easy identification of pipette tip volume
For epTIPS® - the original Eppendorf® 'Totally Integrated Pipetting System' - the tips are optimally coordinated to Eppendorf® pipettes. This results in minimal attachment and ejection forces with the highest level of tightness. epTIPS® pipette tips can be used with pipettes from other manufacturers. Optimal wetting properties, high transparency, and ergonomically optimized cone geometry are a testament to the commitment to product and production quality.
Eppendorf Quality stands for the highest quality and purity achieved by utlizing pure raw materials, autmoated production under cleanroom conditions, quality and functional checks for all lots, and continuous quality assurance throughtout the entire production process. With the strictest control criteria, internally and externally monitored, we guarentee consistent, high-quality pipette tips from lot to lot.
Certifications: Meet EN ISO 8655 requirements.
Packaging: Available as epTIPS® Box (reusable box meant for use with epTIPS® Reloads), as epTIPS® Racks (boxes containing racks of 96, 48, or 24 tips, depending on tip volume), as epTIPS® Reloads (packaged as either dual-sided or stacked racks, depending on tip volume), or as epTIPS® Bulk (packaged in resealable bags).