Would you like to log in again?
Pierce™ Anti-GST Coated Plates, 8-Well Strip, Thermo Scientific
Supplier: Thermo Scientific
Pierce
15144 15144
PI15144EA 307.74 USD InStock
CAPI15144 PI15144
Pierce™ Anti-GST Coated Plates, 8-Well Strip, Thermo Scientific
Microplates Immunology Microplates and Strips
Thermo Scientific Pierce Anti-GST Coated Plates are polystyrene microplates coated with mouse monoclonal anti-GST antibody and pre-blocked for immediate use
  • Detects GST fusion proteins down to 7.5ng/well
  • Binds native or denatured forms of GST; an alternative to glutathione-coated plates
  • Pre-blocked with SuperBlock blocking Buffer
  • Improved signal-to-noise ratio
Store plates at 4 °C and the unused plates can be stored in resealable bag.
Ordering information: Store plates at 4 °C. Store opened, unused plates in a resealable bag with desiccant at 4 °C. Shipped at ambient temperature.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty