Would you like to log in again?
Proteinase K
Supplier: Pierce Chemical
Pierce
17916 17916
PI17916EA 254.58 USD InStock
CAPI17916 PI17916
Proteinase K
Enzymes
Thermo Scientific Pierce Proteinase K is a protease that cleaves at the carboxyl side of aliphatic, aromatic or hydrophobic residues and is commonly used to digest and inactivate DNase and RNase in nucleic acid purification.
Proteinase K is a subtilisin-like endolytic protease that is isolated from the saprophytic fungus Tritirachium album. It has a high activity that is stable across a wide range of pH and temperature conditions and is suited to short digestion times. The activity of proteinase K is increased at elevated temperatures up to 65 ˚C. Calcium is not essential to the function of proteinase K. Therefore, EDTA and other chelating agents do not interfere with the activity and may be used alongside proteinase K to inactivate calcium-dependent nucleases in DNA and RNA preparation.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty