Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N-Dimethylformamide ≥99.5%, Sequencing grade, Pierce™

Supplier: Thermo Scientific
Synonyms: DimethylformamideDMFDMFAFormic acid dimethylamideFormyldimethylamineN-Formyldimethylamine

20673 20673
PI20673EA 90.66 USD InStock
PI20673 CAPI20673
N,N-Dimethylformamide ≥99.5%, Sequencing grade, Pierce™
N,N-Dimethylformamide
Thermo Scientific Pierce Dimethylformamide (DMF) is a sequencing-grade preparation with high purity and consistent absorptivity properties for use in HPLC and mass spectrometry methods.

  • Highly purified and tested to ensure reliable performance in HPLC, mass spectrometry and sequencing applications
  • Packaged in solvent-rinsed amber glass bottles and sealed with PTFE-lined fluorocarbon caps

This sequencing-grade dimethylformamide is specially purified by a proprietary method and tested to ensure lot-to-lot consistency with a low UV absorbance to provide you with the most sensitive detection across all wavelengths. DMF is packaged in solvent-rinsed amber glass bottles and sealed with PTFE-lined fluorocarbon caps for ultimate protection.

Pierce sequencing-grade DMF is specially purified and tested to the highest specifications to ensure the integrity of your data, maximize sensitivity in your assay and to prolong the life of your equipment.

Caution: For Research Use Only. Not for use in diagnostic procedures.


Formula: HCON(CH₃)₂
MW: 73.09 g/mol
Boiling Pt: 153 °C (1013 hPa)
Melting Pt: –61 °C
Density: 0.949 g/cm³ (20 °C)
Flash Pt: 57.5 °C (closed cup)
MDL Number: MFCD00003284
CAS Number: 68-12-2
UN: 2265
ADR: 3,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/25/06/2302506.jpg https://us-legacy.vwr.com/stibo/web/std.lang.all/06/12/16640612.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty