Would you like to log in again?
Pierce™ Streptavidin Poly-HRP, Thermo Scientific
Supplier: Thermo Fisher Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Pierce
21140 21140
PI21140EA 306.94 USD InStock
CAPI21140 PI21140
Pierce™ Streptavidin Poly-HRP, Thermo Scientific
Protein Purification Protein Purification Reagents
Thermo Scientific Pierce Streptavidin Poly-HRP conjugate is biotin-binding protein conjugated with polymers of horseradish peroxidase, enabling signal amplification and detection of biotinylated antibodies for IHC and other methods.
  • Detect low-abundant targets (low picogram-femtogram range) with high signal-to-noise ratios
  • Compatible with chromogenic, fluorogenic and chemiluminescent substrates, also with ELISA, Western blotting and immunohistochemistry (IHC)
  • Can be directly substituted into immunoassays and other detection assays
  • Requires less conjugate per assay and standard HRP conjugates
Streptavidin Poly-HRP conjugate is designed to deliver the highest sensitivity and low background in immunoassays where sample volume is limited or when the target molecule is present at low levels. Streptavidin Poly-HRP is purified to remove unconjugated streptavidin molecules that reduce signal intensity by competing for binding sites with horseradish peroxidase (HRP)-conjugates molecules. In addition, the conjugate is devoid of HRP monomers that can cause background signal. The Pierce Streptavidin Poly-HRP is compatible with chromogenic, fluorogenic and chemiluminescent HRP substrates used in ELISA, Western blotting, immunohistochemistry (IHC) and nucleic acid hybridization assays.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty