Would you like to log in again?
Pierce™ Color Silver Stain Kit, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce™
24597 24597
PI24597EA 401.58 USD InStock
PI24597 CAPI24597
Pierce™ Color Silver Stain Kit, Thermo Scientific
Stains Electrophoresis Stains
Thermo Scientific Pierce Color Silver Stain Kit detects proteins electrophoresed in 1D and 2D polyacrylamide gels using metallic silver to produce color variations that facilitate differentiation of protein bands and spots.
Features of the Color Silver Stain Kit:
Customizable—kit is a reagent set that allows customized formulation of working reagents and staining protocols for particular gel formats and silver-staining applications
Sensitive—detects many proteins at 0.1ng per square millimeter in standard-thickness gels
Broad protein detectability—stains a wider range of protein types than monochromatic silver stains (proteins that do not bind silver are revealed as yellow bands or spots)
Color differentiation—gel-separated proteins and polypeptide-containing macromolecules stain in five basic colors: black, blue, brown, red and yellow
Profiling resolution—multicolored detection of DNA, lipids and polysaccharides in addition to typical proteins provides additional landmarks to aid in 2D gel profiling
The Color Silver Stain Kit contains four reagents and a flexible protocol for staining proteins in 1D and 2D electrophoresis gels. The method requires more time than monochromatic silver stains but provides greater flexibility in adapting the concentration of working solutions to obtain effective staining with different gel types and thicknesses and unusual protein targets. Proteins stain as black, blue, brown, red or yellow bands or spots according to a combination of their specific charge and other characteristics. Once results are known for a particular protein of interest, these slight color differences provide for easy band and spot identification.
Includes:
Four reagent solutions (500 mL each): silver, reducer base, reducer aldehyde and stabilizer base
Ordering information: For Research Use Only. Not for use in diagnostic procedures. Kit contains sufficient reagents to stain 25 (18 cm2) or 40 (130 cm2) gels
500 mL Silver Reagent
500 mL Reducer Aldehyde Reagent
500 mL Reducer Base Reagent
500 mL Stabilizer Base Reagent
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty