Would you like to log in again?
Pierce™ Peroxidase Suppressor, Thermo Scientific
Supplier: Thermo Scientific
Pierce
35000 35000
PI35000EA 224.86 USD InStock
CAPI35000 PI35000
Pierce™ Peroxidase Suppressor, Thermo Scientific
Assays Cellular Assays
Thermo Scientific Pierce Peroxidase Suppressor is a stable, easy-to-use inhibitor of endogenous peroxidase activity from cells and tissues to reduce background signal in HRP-based immunohistochemistry applications.
  • Stable
  • One component
  • Ready-to-use
  • Effectively inhibits peroxidase activity without interfering with antigen-antibody interactions
This peroxidase suppressor is optimized for immunohistochemical applications. The Peroxidase Suppressor is a stable, easy-to-use inhibitor of endogenous peroxidase activity. This proprietary formulation is the most effective peroxidase inhibitor available. The immunoperoxidase method is a commonly used detection method for immunostaining of cells and tissues. However, the analysis often is complicated by the presence of endogenous peroxidase and 'pseudoperoxidase' activity in the cells and tissues. This presence results in nonspecific staining when a substrate such as diaminobenzidine (DAB) is present. Nonspecific staining is difficult to distinguish from the immunochemical marker, making it difficult to distinguish the final location of the antigen of interest. The typical method of inhibiting endogenous peroxidase activity involves hydrogen peroxide or hydrogen peroxide in methanol. The peroxidase suppressor inhibits peroxidase activity even more effectively than the method using hydrogen peroxide in methanol. Plus, the Peroxidase Suppressor does not affect antibody activity (see figure). Peroxidase Suppressor also can be used on blots of whole cell supernatant prior to the addition of the horseradish peroxidase (HRP) marker.
Caution: Upon receipt store at 4°C. For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty