Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Pierce™ FITC Antibody Labeling Kit, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
53027 53027
PI53027EA 563.45 USD InStock
PI53027 CAPI53027
Pierce™ FITC Antibody Labeling Kit, Thermo Scientific
Protein Linking and Labeling Systems
Pierce™ FITC Antibody Labeling Kit is a multi-component kit containing a high performance derivative of fluorescein dye, activated for easy and reliable labeling of antibodies, proteins and other molecules for use as fluorescent probes.
  • Convenient, easy to use kit with FITC to label and purify antibody in about one hour
  • Amine-specific labeling – the isothiocyanate variety of fluorescein efficiently labels antibodies and other purified proteins at primary amines
  • Optimised kit protocol results in antibodies with excellent dye protein ratios for optimum activity and fluorescence
  • Single-use vials of reagent for added convenience
  • Purification resin and easy-to-use spin columns, ensuring rapid and efficient removal of non-reacted dye and excellent protein recovery
Delivery information: Standard kit includes FITC (3×50 µg vials), borate buffer (1 mL), spin columns (6 each), collection tubes (12 each) and purification resin (5 mL).
Caution: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty