Would you like to log in again?
myImageAnalysis™ Software, Thermo Scientific
Supplier: Thermo Scientific
62237
PI62237EA 2979.31 USD InStock
PI62237
myImageAnalysis™ Software, Thermo Scientific
Photodocumentation Systems
Thermo Scientific myImageAnalysis Software is a full-featured, multifunction computer program to analyze and edit digital images of electrophoresis gels and blots that were acquired using gel documentation imagers or scanners.
Ordering information: For Research Use Only. Not for use in diagnostic procedures. Contents: 1 software installation CD
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty