Would you like to log in again?
Green Filter for myECL™ Imager, Thermo Scientific
Supplier: Thermo Scientific
62239
PI62239EA 1567.85 USD InStock
PI62239
Green Filter for myECL™ Imager, Thermo Scientific
Photodocumentation Systems
The Thermo Scientific 510-550nm bandpass filter is a threaded, 50mm-diameter glass filter
This product is used for the addition of a green filter to one of two open positions in the myECL Imager for detection (emission) wavelengths of common fluorescent nucleic acid stains, such as SYBR* Green Nucleic Acid Gel Stain and SYBR Safe DNA Gel Stain.
Ordering information: For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty