Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Pierce™ Graphite Spin Columns, Thermo Scientific
Supplier: Thermo Scientific
Pierce™
88302 88302
PI88302EA 237.78 USD InStock
CAPI88302 PI88302
Pierce™ Graphite Spin Columns, Thermo Scientific
Filters Centrifugal Filters
These Graphite spin columns improve phosphopeptide analysis by efficiently binding hydrophilic peptides and efficiently removing urea, salts and other contaminants before mass spectrometry analysis.
  • Convenient spin format for parallel processing of multiple samples
  • High-binding capacity with excellent recovery of up to 100 µg of hydrophilic peptides per column
  • Porous graphite resin for cleaning up phosphopeptide samples before ms analysis
These Graphite spin columns enable fast and efficient capture, concentration, desalting and elution of hydrophilic peptides in less than 10 minutes. These columns are ideal for improving mass spectrometric (MS) analyses of samples from protein digests), strong-cation exchange fractions, and enriched phosphopeptides eluted from titanium dioxide and immobilized metal affinity chromatography (IMAC) columns and tips.
Ordering information: Store at room temperature.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty