Would you like to log in again?
Bovine Serum Albumin (BSA)
Supplier: Pierce Chemical
88341
PI88341EA 195.69 USD InStock
PI88341
Bovine Serum Albumin (BSA)
Proteins and Peptides
The Thermo Scientific™ Pierce™ BSA Protein Digest Standard, LC-MS Grade, is a validated protein digest optimized for use as a quality control sample for liquid chromatography (LC) and mass spectrometry (MS) analysis of proteomic samples.Positive control sample—BSA digest optimized and validated as a quality control standard for mass spectrometry applicationsExcellent sequence coverage—greater than 85% sequence coverageValidated peptide quality—digestion procedure optimized for minimal missed cleavages and over-alkylationRigorously tested—high-quality, consistent BSA protein digest documented via lot-specific certificates of analysisStable—provided in a stable lyophilized formatThe Pierce BSA Protein Digest Standard is produced using high quality bovine serum albumin (BSA) and mass spectrometry-grade trypsin. The BSA is fully reduced, alkylated (iodoacetamide), and desalted (C18) after trypsin digestion to provide a quality LC/MS-grade standard that is free of intact protein. The peptide digest standard provides excellent sequence coverage with minimal over-alkylation and missed cleavages. This digest standard has been robustly tested for peptide quality, digestion efficiency, and lot-to-lot uniformity and is suitable for LC and LC-MS applications.
For Research Use Only. Not for use in diagnostic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty