Would you like to log in again?
TC Ryder Needle Holders, OR Grade, Sklar
Supplier: Sklar
21-8014 21-8013 21-8019 21-8016 21-8021 21-8020 21-8012 21-8022
89023-328EA 445.29 USD InStock
89023-328 89023-336 89023-338 89023-342 89023-332 89023-334 89023-340 89023-330
TC Ryder Needle Holders, OR Grade, Sklar
Forceps
TC Ryder Needle Holders are a ratcheted, finger ring instrument, that features narrow jaws, used with small suture needles in cardiovascular, plastic, and neurosurgical procedures.
  • Censitrac Ready products are industry standard and constructed of stainless steel
  • Latex-free
Tungsten carbide micro cross-serrated tips grip the small needle and assist with keeping it from twisting during suturing.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty