Would you like to log in again?
Aufricht Glabellar Rasp, OR Grade, Sklar
Supplier: Sklar
40-9840
10196-598EA 292.03 USD InStock
10196-598
Aufricht Glabellar Rasp, OR Grade, Sklar
Raspatories
Aufricht Glabellar Raspatory is a debulking instrument used for filing rough bone edges in head and neck procedures.
This product has coarse, horizontal serrations and a slightly curved tip intended for forward cutting.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty