Would you like to log in again?
Joseph Raspatory, OR Grade, Sklar
Supplier: Sklar
40-9825
10196-596EA 232.46 USD InStock
10196-596
Joseph Raspatory, OR Grade, Sklar
Raspatories
Joseph Raspatory is a debulking instrument used in ENT procedures and to smooth and debulk other small bones or joints.
  • Non sterile, latex free and reusable
This instrument is a single end raspatory that has fine cross-serrations across a flat, oval shaped blade.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty