Would you like to log in again?
TC Mayo-Hegar Needle Holder, OR Grade, Sklar®
Supplier: Sklar
21-8035 21-8111 21-2914 21-8112 21-8113 21-8031 21-8051 21-8034 21-8033 21-8030 21-8032
76192-770EA 307.39 USD InStock
76192-770 76192-716 89023-358 89023-364 76192-768 89023-366 76192-766 76192-348 89023-356 89023-360 89023-362
TC Mayo-Hegar Needle Holder, OR Grade, Sklar®
Forceps
TC Mayo-Hegar Needle Holder is a ratcheted, finger ring instrument available in a wide range of lengths for multiple types of surgical procedures.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Due to it's versatility, this is one of the most commonly used surgical tools for both human and animal procdures. Shorter length products may be used in oral, gynecological, or dermatological procedures, as well as with small animals. Larger lengths may be used for deep tissue suturing such as in bariatric procedures, or in large animal procedures. The Mayo-Hegar needle holder is commonly used with larger, heavier suture needles because of it's wide jaws. Tungsten carbide inserts also maximize control and precision.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty