Would you like to log in again?
TC Stratte Needle Holder, OR Grade, Sklar
Supplier: Sklar
21-8065
89023-396EA 647.41 USD InStock
89023-396
TC Stratte Needle Holder, OR Grade, Sklar
Forceps
Sklar's® tungsten carbide Stratte needle holder is a ratcheted instrument with curved, half serrated jaws ensuring a firm grip on the needle. They are designed for use with 2-0 and 3-0 sutures.
TC instruments are designed to stay sharp for a significantly longer period of time than other standard instruments
TC Stratte Needle Holder and Censitrac Ready products are industry standard and constructed of stainless steel.
Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty