Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Murphy Intestinal Needles, OR Grade, Sklar
Supplier: Sklar
93-1720-3 93-1720-4 93-1720-2 93-1720-1
10185-214PK 29.65 USD InStock
10185-214 10185-212 10185-218 10185-216
Murphy Intestinal Needles, OR Grade, Sklar
Dissection Needles
Murphy Intestinal Needles are used for intestinal anatomosis in veterinary procedures
This product is a #1 needle with a 1/2 circle curvature and a taper point.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty