Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Mayo Cat Gut Needles, OR Grade, Sklar
Supplier: Sklar
93-1718-5 93-1718-7 93-1718-1 93-1718-6 93-1718-2 93-1718-3 93-1718-4
10185-210PK 29.65 USD InStock
10185-210 10185-412 10185-410 10185-416 10185-414 10185-408 10185-406
Mayo Cat Gut Needles, OR Grade, Sklar
Dissection Needles
Mayo Cat Gut Needles are used with what is referred to as catgut sutures
They feature half circle curvature.
The absorbable suture material was made from the intestines or tendons of cattle, sheep, or goats. Most catgut type sutures are made from synthetic materials that are still absorbable by the body. This product is for use in veterinary procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty