Would you like to log in again?
Webster Needle Holders, OR Grade, Sklar
Supplier: Sklar
20-3090
10195-168EA 217.33 USD InStock
10195-168
Webster Needle Holders, OR Grade, Sklar
Forceps
Webster Needle Holders are used with small needles and fine suture material, in plastic surgery or subcutaneous procedures around the face and hands.
They are available with smooth or serrated jaws. Smooth jaws will cause the least damage to the needle, however serrated jaws hold the needle more tightly. A Webster needle holder should not be used with large needles because of the risk of damage to the jaws, thus enabling the instrument insufficient for future use with the intended small needle.
Sklar bonds tungsten carbide inserts to this instrument, which maximizes control and precision. Physician preference determines the type of working surface utilized.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty