Would you like to log in again?
Sklar Black™ Mayo-Hegar Needle Holder, OR Grade, Sklar
Supplier: Sklar
Sklar Black™
90-2288
10199-104EA 373.06 USD InStock
10199-104
Sklar Black™ Mayo-Hegar Needle Holder, OR Grade, Sklar
Forceps
Sklar Black™ Mayo-Hegar Needle Holders are widely used in general surgery for holding medium to large, heavy suture needles.
The heavy jaws have cross-serrations and a groove. This instrument is available in several lengths. The shorter length instrument may be used for procedures such as suturing the uterine cervix, while longer lengths may be used for deep tissue suturing.
Sklar Black™ Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff.
Certifications: All Sklar Black™ coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements. They also comply with the Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79. Sklar Black™ laser coatings are guaranteed for one year against manufacturing defects.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty