Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium sulfate ≥99.0%, crystallised ACS, VWR Chemicals BDH®

Supplier: VWR International

BDHVBDH4618-500G
BDH4618-500GCS 1375.65 USD InStock
BDH4618-500G
Potassium sulfate ≥99.0%, crystallised ACS, VWR Chemicals BDH®
Potassium sulphate


Formula: K₂SO₄
MW: 174.26 g/mol
Boiling Pt: 1698 °C (1013 hPa)
Melting Pt: 1069 °C
Density: 2.66 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011388
CAS Number: 7778-80-5
EINECS: 231-915-5
Merck Index: 12,07845
Order Now
Specification Test Results
Assay Min. 99.0 %
Appearance Crystals/crystall. powder
Appearance (color) White
pH (25°C; 5 %) 5.5 - 8.5
Heavy metals (as Pb) Max. 5 ppm
Insoluble matter Max. 0.01 %
Nitrogen compounds (as N) Max. 5 ppm
Cl (Chloride) Max. 0.001 %
Ca (Calcium) Max. 0.01 %
Fe (Iron) Max. 5 ppm
Mg (Magnesium) Max. 0.005 %
Na (Sodium) Max. 0.02 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty