Would you like to log in again?
Sklarlite XD™ Hartmann Mosquito Forceps, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Sklarlite XD™
23-2501
10193-404EA 210.48 USD InStock
10193-404
Sklarlite XD™ Hartmann Mosquito Forceps, OR Grade, Sklar
Forceps
Sklarlite XD™ Hartmann Mosquito Forceps are used as both a hemostat and for fine tissue dissection in shallow wounds and procedures
Known for their fine tips, short, and fully serrated jaws, these Mosquito Forceps can be used as a clamping instrument to halt blood flow so that cauterization or ligation can be performed. They are also used for holding small, fine sutures. Hartmann Mosquito Forceps are shorter and heavier than Halstead Mosquito Forceps.
Sklarlite XD™ instruments are noticeably lighter and delicate than standard instruments which allow the surgeon the highest degree of precision with the least amount of fatigue. These instruments have been manufactured to be one of the lightest, most delicate general instruments available today.
For plastic and vascular surgeries, Hartmann Mosquito Forceps can be used in pediatric and ophthalmologic procedures.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty