Would you like to log in again?
Sklarlite™ Hartmann Mosquito Forceps, OR Grade, Sklar
Supplier: Sklar
Sklarlite™
23-2100
10193-406EA 174.94 USD InStock
10193-406
Sklarlite™ Hartmann Mosquito Forceps, OR Grade, Sklar
Forceps
Sklarlite™ Hartmann Mosquito forceps are a lighter version of standard mosquito forceps
Known for their fine tips, short, and fully serrated jaws, these Mosquito Forceps can be used as a clamping instrument to halt blood flow, so that cauterization or ligation can be performed. They are used for holding small, fine sutures. Hartmann Mosquito Forceps are shorter and heavier than Halstead Mosquito Forceps.
This lightweight modification provides the surgeon with greater control and less fatigue. Mosquito forceps are used as both a hemostat and for fine tissue dissection in shallow wounds and procedures. For plastic and vascular surgeries, Hartmann Mosquito Forceps can be used in pediatric and ophthalmologic procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty