Would you like to log in again?
Sklarlite™ Halsted Mosquito Forceps, OR Grade, Sklar
Supplier: Sklar
Sklarlite™
23-2105
10194-348EA 133.37 USD InStock
10194-348
Sklarlite™ Halsted Mosquito Forceps, OR Grade, Sklar
Forceps
Sklarlite™ Halsted Mosquito Forceps are a lighter version of standard Sklar mosquito forceps
The ratcheted finger ring handle provides a secure, locking grip to halt blood flow. They are not appropriate for use with large vessels or arteries. A toothed variation is available for grasping delicate tissue in skin grafting, biopsies, or ophthalmologic procedures. Mosquito forceps are known for their fine tips, and short, fully serrated jaws. Halsted Mosquito Forceps are very similar to Hartmann Mosquito Forceps; however, the Hartmann style is finer and more delicate.
This lightweight modification provides the surgeon with greater control and less fatigue. They are used in multiple procedures as a hemostat for small blood vessels.
Ordering information: Available curved or straight, and with a length of 5 1/2".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty