Would you like to log in again?
RPMI 1640, Himedia
Supplier: HiMedia Laboratories
AT162-1L
10770-348EA 25.83 USD InStock
10770-348
RPMI 1640, Himedia
Cell Culture Media
Roswell Park Memorial Institute (RPMI) media are a series of media for the culture of human normal and neoplastic cells in vitro.
  • It needs to be supplemented with 5-20% fetal bovine serum
  • They also known for supporting growth of cells in the absence of serum
RPMI-1640 is the most commonly used medium in the series. The medium is extensively used for a wide range of anchorage dependant cell lines.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty