Would you like to log in again?
Sklarlite™ Lister Bandage Scissors, OR Grade, Sklar
Supplier: Sklar
Sklarlite™
23-1199 23-1200
10195-116EA 149.04 USD InStock
10195-116 10195-118
Sklarlite™ Lister Bandage Scissors, OR Grade, Sklar
Scissors
Sklarlite™ Lister Bandage Scissors are a versatile instrument with multiple uses
The jaws of the Lister Bandage Scissors are angled with the lower blade being slightly longer. The tip of the lower blade features a flattened blunt nodule that is intended to slide between bandages and skin without harming the skin. This feature allows Lister Bandage Scissors to occasionally be used during cesarean section to open the uterus without causing inadvertent injury to the baby. Ring handles can be equal in size or with one ring larger.
Sklarlite™ instruments are a lighter version of standard Sklar instruments. A manufacturing process developed by Sklar shaves microns off standard and delicate pattern instruments while maintaining structural integrity and durability. This lightweight modification provides the surgeon with greater control and less fatigue.
They are used for sizing dressings and removing circumferential bandages. They may be used to cut tougher material such as plaster, fabric and occasionally umbilical cord.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty