Would you like to log in again?
Normal Goat Serum, Jackson Immuno
Supplier: Jackson Immunoresearch Laboratories
005-000-121
102643-594EA 78.77 USD InStock
102643-594
Normal Goat Serum, Jackson Immuno
Sera (Serum)
Normal serums are lipid extracted to improve clarity, dialyzed against phosphate buffered saline (PBS) containing sodium azide, and freeze dried.
  • Lipid extracted to improve clarity
  • Recommended blocking agent
  • Dialyzed against phosphate buffered saline containing sodium azide
Normal serum diluted to 5% (v/v) in PBS is strongly recommended as a blocking reagent to reduce background from non-specific, conserved sequence and/or Fc receptor binding. Best results are obtained with diluted normal serum from the same host as the labeled antibody, as a separate incubation step before addition of the primary antibody.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty