Would you like to log in again?
Mini Tischler Micro-Cut Universal Style Biopsy Punch, OR Grade, Sklar
Supplier: Sklar
90-8964
10184-196EA 1958.18 USD InStock
10184-196
Mini Tischler Micro-Cut Universal Style Biopsy Punch, OR Grade, Sklar
Biopsy Punches
Sklar's Mini Tischler Micro-Cut Universal Style Biopsy Punch Forceps are an OB/GYN instrument most commonly used in colposcopy procedures to obtain cervical tissue samples for biopsy
The mini Tischler bite, 2.3mm x 4.2mm, is slightly smaller than a standard Tischler bite and takes a smaller tissue sample. The mini or baby style biopsy punch is often preferred because there is typically less bleeding and pain when the smaller tissue sample is removed. This product is a complete instrument which consists of a handle and a detachable cannula/tip so other universal tip styles may be used with one handle.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty