Would you like to log in again?
Surgi-OR™ TC Webster Needle Holders, Mid Grade, Sklar
Supplier: Sklar
Surgi-OR™
95-809
89023-724EA 48.76 USD InStock
89023-724
Surgi-OR™ TC Webster Needle Holders, Mid Grade, Sklar
Forceps
Surgi-OR™ TC Webster Needle Holders are used with small needles and fine suture material, commonly in plastic surgery or subcutaneous procedures.
They are available with smooth or serrated jaws. Smooth jaws will cause the least damage to the needle; however, serrated jaws hold the needle more tightly. Physician preference typically determines the type of working surface utilized.
Hand-crafted in Pakistan, Surgi-OR™ TC mid-grade instruments are forged from the highest quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers. They are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty