Would you like to log in again?
Econo™ Sterile Crile-Wood Needle Holder, Sklar
Supplier: Sklar
Econo™
96-2590 96-2591
89023-922CS 496.46 USD InStock
89023-922 89023-924
Econo™ Sterile Crile-Wood Needle Holder, Sklar
Forceps
Econo™ Sterile Crile-Wood Needle Holder is used to hold and guide small to medium size needles and suture materials
The ratcheted finger ring handle aids in securing needles for both continuous and interrupted closures.
Econo™ Sterile instruments are ready-to-use, sterile, disposable, floor-grade instruments made in Pakistan. Econo™ Sterile instruments eliminate the need for cleaning and reprocessing. Because they are both sterile and disposable, Econo™ Sterile instruments reduce the likelihood of cross-contamination.
The Crile-Wood Needle Holder has a tapered, blunt tip and is more delicate than the Mayo-Hegar but slightly less delicate than Webster, Halsey, or DeBakey needle holders.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty