Would you like to log in again?
Plastic Bottles, Narrow Mouth, HDPE, Natural, C&G Containers
Supplier: C&G Containers
LPN128900000 LPN128200014 LPN128200010
71001-005CS 254.46 USD InStock
71001-005 71001-006 71001-009
Plastic Bottles, Narrow Mouth, HDPE, Natural, C&G Containers
Bottles Plastic Bottles
These plastic bottles are used for the collection of aqueous media for testing metals and a variety of inorganic parameters, including alkalinity, anions, acidity, cyanide, sulfate, and hardness.
Pre-cleaned and Certified products contain the certificate of analysis and Cleaning Protocol Form in the product case.
A lot certificate of analysis is generated with the analytical results obtained, including the concentration reporting limits and units of measurement, for Pre-cleaned and Certified products. This certificate of analysis is included in each product case.
Wash protocols for these bottles include; A1 (nitric acid wash), B1 (deionized water wash), C1 (hydrochloric acid wash), and L3 (no wash).
Not all analytical parameters are included on the certificate of analysis. Please refer to the Certificate of Analysis for certified analytes and corresponding reporting limits. Secondary uses for these containers include containment and archiving.
Ordering information: Container measurements and capacity 'contain' volumes may vary.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty