Would you like to log in again?
Micro Tubes with Looped Screw Cap, Type A, Sarstedt
Supplier: Sarstedt
72.730.105 72.730.106 72.730.107 72.730.100
101093-764PK 183.92 USD InStock
101093-764 101093-755 101093-608 101093-760
Micro Tubes with Looped Screw Cap, Type A, Sarstedt
Tubes Microtubes
Type A micro tubes are designed for transport, storage, and sample processing in the laboratory.
  • Leak-proof looped screw cap with O-ring
  • Safe identification of screw cap/tube
  • Autoclavable at 121 °C
  • Knurls enable single-handed operation in special Sarstedt racks for micro tubes
The neutral looped screw cap can be provided with inserts for color coding.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty