Would you like to log in again?
Schumacher-Van Doren Biopsy Punch, OR Grade, Sklar
Supplier: Sklar
90-7511
10193-996EA 1107.75 USD InStock
10193-996
Schumacher-Van Doren Biopsy Punch, OR Grade, Sklar
Biopsy Punches
Schumacher-Van Doren Biopsy Punch is a non-ratcheted, finger ring instrument used for removing uterine tissue samples for biopsy.
Long shanks curve slightly after the handle so that the users hands may be lower and do not obstruct the view of the surgical site. The short, gently curved jaws are easily managed and the bite provides a 7 x 7 x 4-mm triangular tissue sample.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty